accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112811-71-9

IUPAC Name: ethyl 1-cyclopropyl-6,7-difluoro-8-methoxy-4-oxo-1,4-dihydroquinoline-3-carboxylate
Synonyms: ethyl 1-cyclopropyl-6,7-difluoro-8-methoxy-4-oxoquinoline-3-carboxylate
Molecular Weight (g/mol): 323.3
Molecular Formula: C16H15F2NO4
InChi Key: XPAOPAPDCRLMTR-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CN(C2CC2)C2=C(OC)C(F)=C(F)C=C2C1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results