accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112881-51-3

IUPAC Name: 2,6-bis(pyridin-2-yl)-4,4'-bipyridine
Synonyms: 2,6-bis(pyridin-2-yl)-4,4'-bipyridine
Molecular Weight (g/mol): 310.36
Molecular Formula: C20H14N4
InChi Key: DPPKPCLKZOLTMB-UHFFFAOYSA-N
SMILES: C1=CC=C(N=C1)C1=CC(=CC(=N1)C1=CC=CC=N1)C1=CC=NC=C1
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results