accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CS(R)OHONHOO

CAS RN 112883-40-6

IUPAC Name: (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-4-(methylsulfanyl)butanoic acid
Synonyms: (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-4-(methylsulfanyl)butanoic acid
Molecular Weight (g/mol): 371.45
Molecular Formula: C20H21NO4S
InChi Key: BUBGAUHBELNDEW-GOSISDBHSA-N
SMILES: CSCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)