accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 112883-43-9

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(naphthalen-2-yl)propanoate
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(naphthalen-2-yl)propanoate
Molecular Weight (g/mol): 436.49
Molecular Formula: C28H22NO4
InChi Key: JYUTZJVERLGMQZ-SANMLTNESA-M
SMILES: [O-]C(=O)[C@H](CC1=CC=C2C=CC=CC2=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (4)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)