accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1129-28-8

IUPAC Name: methyl 3-(bromomethyl)benzoate
Synonyms: methyl 3-(bromomethyl)benzoate
Molecular Weight (g/mol): 229.07
Molecular Formula: C9H9BrO2
InChi Key: YUHSMQQNPRLEEJ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC=CC(CBr)=C1
Boiling Point 
  • (1)
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (2)
  • (5)
  • (1)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)