accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNHOOH3C

CAS RN 1129-41-5

IUPAC Name: 3-methylphenyl N-methylcarbamate
Synonyms: kumiai tsumaunka MTMC tsumacide dicresyl N-methylcarbamate N-methyl-M-tolylcarbamate metholcarb metacrate metolcarb
Molecular Weight (g/mol): 165.19
Molecular Formula: C9H11NO2
InChi Key: VOEYXMAFNDNNED-UHFFFAOYSA-N
SMILES: CNC(=O)OC1=CC=CC(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results