accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1129540-65-3

IUPAC Name: 2-bromo-6-(morpholin-4-yl)benzonitrile
Synonyms: 2-bromo-6-(morpholin-4-yl)benzonitrile
Molecular Weight (g/mol): 267.13
Molecular Formula: C11H11BrN2O
InChi Key: HKIGGSMIIBGCKZ-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(N2CCOCC2)=C1C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results