accessibility menu, dialog, popup

UserName

Viewing profile as user:
NCSFF

CAS RN 113028-75-4

IUPAC Name: 1,2-difluoro-4-isothiocyanatobenzene
Synonyms: 1,2-difluoro-4-isothiocyanatobenzene
Molecular Weight (g/mol): 171.16
Molecular Formula: C7H3F2NS
InChi Key: SVZKYXSJICYUOH-UHFFFAOYSA-N
SMILES: FC1=C(F)C=C(C=C1)N=C=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results