accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1131-09-5

IUPAC Name: 2-(1-benzothiophen-3-yl)acetic acid
Synonyms: 1-benzothiophen-3-ylacetic acid
Molecular Weight (g/mol): 192.23
Molecular Formula: C10H8O2S
InChi Key: VFZQJKXVHYZXMM-UHFFFAOYSA-N
SMILES: OC(=O)CC1=CSC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results