accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOO

CAS RN 1131-15-3

IUPAC Name: 3-phenyloxolane-2,5-dione
Synonyms: 3-phenyloxolane-2,5-dione
Molecular Weight (g/mol): 176.17
Molecular Formula: C10H8O3
InChi Key: HDFKMLFDDYWABF-UHFFFAOYNA-N
SMILES: O=C1CC(C(=O)O1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results