accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1131-60-8

IUPAC Name: 4-cyclohexylphenol
Synonyms: P-cyclohexylphenol
Molecular Weight (g/mol): 176.26
Molecular Formula: C12H16O
InChi Key: OAHMVZYHIJQTQC-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1CCCCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results