accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1131-62-0

IUPAC Name: 1-(3,4-dimethoxyphenyl)ethan-1-one
Synonyms: 1-(3,4-dimethoxyphenyl)ethanone
Molecular Weight (g/mol): 180.2
Molecular Formula: C10H12O3
InChi Key: IQZLUWLMQNGTIW-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1OC)C(C)=O
Boiling Point 
  • (3)
  • (2)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (5)
Percent Purity 
  • (2)
  • (3)
  • (5)
Quantity 
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (2)
  • (3)
  • (5)
  • (5)
  • (2)
  • (3)
  • (5)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (6)