accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 113100-53-1

IUPAC Name: 1-methyl-3-(trifluoromethyl)-1H-pyrazole-4-carboxylate
Synonyms: 1-methyl-3-(trifluoromethyl)pyrazole-4-carboxylate
Molecular Weight (g/mol): 193.11
Molecular Formula: C6H4F3N2O2
InChi Key: FZNKJQNEJGXCJH-UHFFFAOYSA-M
SMILES: CN1C=C(C([O-])=O)C(=N1)C(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results