accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 113118-81-3

IUPAC Name: 5-bromopyridine-3-carbaldehyde
Synonyms: 5-bromopyridine-3-carbaldehyde
Molecular Weight (g/mol): 186.01
Molecular Formula: C6H4BrNO
InChi Key: NGUVGKAEOFPLDT-UHFFFAOYSA-N
SMILES: BrC1=CN=CC(C=O)=C1
Molecular Weight (g/mol) 
  • (4)
  • (3)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (4)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (2)
  • (5)
  • (4)
  • (3)
  • (4)