accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 113170-55-1

IUPAC Name: bis({[5-(1,2-dihydroxyethyl)-4-hydroxy-2-oxo-2,5-dihydrofuran-3-yl]oxy}phosphonic acid) trimagnesium
Synonyms: bis([5-(1,2-dihydroxyethyl)-4-hydroxy-2-oxo-5H-furan-3-yl]oxyphosphonic acid) trimagnesium
Molecular Weight (g/mol): 585.12
Molecular Formula: C12H18Mg3O18P2
InChi Key: IRBRZNUQJUHOBT-UHFFFAOYNA-N
SMILES: [Mg].[Mg].[Mg].OCC(O)C1OC(=O)C(OP(O)(O)=O)=C1O.OCC(O)C1OC(=O)C(OP(O)(O)=O)=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results