accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CO

CAS RN 1131912-76-9

IUPAC Name: 4,4,5,5-tetramethyl-2-(oxan-4-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(oxan-4-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 212.1
Molecular Formula: C11H21BO3
InChi Key: NLSMOSUUBUCSPL-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1CCOCC1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)