CAS RN 1132-26-9
IUPAC Name:
2-amino-3-phenylbutanoic acid
Synonyms:
2-amino-3-phenylbutanoic acid
Molecular Weight (g/mol):
179.22
Molecular Formula:
C10H13NO2
InChi Key:
IRZQDMYEJPNDEN-UHFFFAOYNA-N
SMILES:
CC(C(N)C(O)=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Sigma Aldrich D-Allose
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.