accessibility menu, dialog, popup

UserName

Viewing profile as user:
Se

CAS RN 1132-39-4

IUPAC Name: (phenylselanyl)benzene
Synonyms: diphenyl selenide
Molecular Weight (g/mol): 233.18
Molecular Formula: C12H10Se
InChi Key: ORQWTLCYLDRDHK-UHFFFAOYSA-N
SMILES: [Se](C1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (1)
  • (3)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)