accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1132-95-2

IUPAC Name: 1,1-bis(propan-2-yloxy)cyclohexane
Synonyms: 1,1-diisopropoxycyclohexane
Molecular Weight (g/mol): 200.32
Molecular Formula: C12H24O2
InChi Key: PLNTYOACSMHWBN-UHFFFAOYSA-N
SMILES: CC(C)OC1(CCCCC1)OC(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results