accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOFNH2NH2

CAS RN 113269-06-0

IUPAC Name: 4-fluoro-5-nitrobenzene-1,2-diamine
Synonyms: 4-fluoro-5-nitrobenzene-1,2-diamine
Molecular Weight (g/mol): 171.13
Molecular Formula: C6H6FN3O2
InChi Key: QZNALIMEKAACKP-UHFFFAOYSA-N
SMILES: NC1=CC(F)=C(C=C1N)[N+]([O-])=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)