accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1133-77-3

IUPAC Name: 1-phenyl-1H-pyrazole-5-carboxylic acid
Synonyms: 2-phenylpyrazole-3-carboxylic acid
Molecular Weight (g/mol): 188.19
Molecular Formula: C10H8N2O2
InChi Key: FLYDUXCFCARXHI-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=NN1C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results