accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSiH3CCH3OOONNN

CAS RN 113306-55-1

IUPAC Name: 1H-1,2,3-benzotriazol-1-yl 2-(trimethylsilyl)ethyl carbonate
Synonyms: 1,2,3-benzotriazol-1-yl 2-(trimethylsilyl)ethyl carbonate
Molecular Weight (g/mol): 279.37
Molecular Formula: C12H17N3O3Si
InChi Key: BJUJSLXVBATBSB-UHFFFAOYSA-N
SMILES: C[Si](C)(C)CCOC(=O)ON1N=NC2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results