accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CSONCH3

CAS RN 1134-23-2

IUPAC Name: N-cyclohexyl-N-ethyl(ethylsulfanyl)formamide
Synonyms: cycloate
Molecular Weight (g/mol): 215.36
Molecular Formula: C11H21NOS
InChi Key: DFCAFRGABIXSDS-UHFFFAOYSA-N
SMILES: CCSC(=O)N(CC)C1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results