accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1134-35-6

IUPAC Name: 4,4'-dimethyl-2,2'-bipyridine
Synonyms: 4,4'-dimethyl-2,2'-bipyridine
Molecular Weight (g/mol): 184.24
Molecular Formula: C12H12N2
InChi Key: NBPGPQJFYXNFKN-UHFFFAOYSA-N
SMILES: CC1=CC=NC(=C1)C1=CC(C)=CC=N1
Molecular Weight (g/mol) 
  • (6)
  • (6)
Percent Purity 
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
Quantity 
  • (4)
  • (4)
  • (4)
Physical Form 
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (3)
  • (6)
  • (6)
  • (6)
  • (3)
  • (3)
  • (2)
  • (3)
  • (1)
  • (4)
  • (4)
  • (4)
  • (6)