accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1134-94-7

IUPAC Name: 2-(phenylsulfanyl)aniline
Synonyms: 2-(phenylsulfanyl)aniline
Molecular Weight (g/mol): 201.29
Molecular Formula: C12H11NS
InChi Key: DGBISJKLNVVJGD-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1SC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)