accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHFFNOO

CAS RN 113512-57-5

IUPAC Name: 2,4-difluoro-5-nitrophenol
Synonyms: 2,4-difluoro-5-nitrophenol
Molecular Weight (g/mol): 175.09
Molecular Formula: C6H3F2NO3
InChi Key: SMRYCTJAGPDVEH-UHFFFAOYSA-N
SMILES: OC1=C(F)C=C(F)C(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results