CAS RN 1135389-59-1
IUPAC Name:
(1,3-dioxolan-2-yl)methanide; bromozincylium
Synonyms:
1,3-dioxolan-2-ylmethanide; bromozincylium
Molecular Weight (g/mol):
232.38
Molecular Formula:
C4H7BrO2Zn
InChi Key:
ZEBRGOCCZOTNDU-UHFFFAOYSA-M
SMILES:
[Zn+]Br.[CH2-]C1OCCO1
1
–
1
of
1
results
1
Promotions Available
(1,3-Dioxolan-2-ylmethyl)zinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
(1,3-Dioxolan-2-ylmethyl)zinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles
CAS: 1135389-59-1 Molecular Formula: C4H7BrO2Zn Molecular Weight (g/mol): 232.382 MDL Number: MFCD07698709 InChI Key: ZEBRGOCCZOTNDU-UHFFFAOYSA-M Synonym: bromo 1,3-dioxolan-2-ylmethyl zinc,1,3-dioxolan-2-ylmethyl zinc bromide,lojrlqrgbkthdb-uhfffaoysa-m,1,3-dioxolan-2-ylmethyl zinc bromide, 0.6 m in thf,1,3-dioxolan-2-ylmethyl zinc bromide, 0.50 m in thf,1,3-dioxolan-2-ylmethyl zinc bromide 0.5 m in tetrahydrofuran,1,3-dioxolan-2-ylmethyl zinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles PubChem CID: 24722674 IUPAC Name: bromozinc(1+);2-methanidyl-1,3-dioxolane SMILES: [CH2-]C1OCCO1.[Zn+]Br