accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOHOOOOOO

CAS RN 113544-59-5

IUPAC Name: [3,4,5-tris(benzoyloxy)-6-hydroxyoxan-2-yl]methyl benzoate
Synonyms: [3,4,5-tris(benzoyloxy)-6-hydroxyoxan-2-yl]methyl benzoate
Molecular Weight (g/mol): 596.59
Molecular Formula: C34H28O10
InChi Key: FCDYAJBVISGNLC-UHFFFAOYNA-N
SMILES: OC1OC(COC(=O)C2=CC=CC=C2)C(OC(=O)C2=CC=CC=C2)C(OC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results