accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNF

CAS RN 113770-88-0

IUPAC Name: 3-fluoropyridine-4-carbonitrile
Synonyms: 3-fluoropyridine-4-carbonitrile
Molecular Weight (g/mol): 122.1
Molecular Formula: C6H3FN2
InChi Key: RFQHDTRSEWCXJP-UHFFFAOYSA-N
SMILES: FC1=C(C=CN=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results