accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 113798-74-6

IUPAC Name: 2,3,6-trifluorophenol
Synonyms: 2,3,6-trifluorophenol
Molecular Weight (g/mol): 148.08
Molecular Formula: C6H3F3O
InChi Key: QSFGUSFDWCVXNR-UHFFFAOYSA-N
SMILES: OC1=C(F)C=CC(F)=C1F
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)