accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1138-56-3

IUPAC Name: 4-butoxybenzene-1-sulfonyl chloride
Synonyms: 4-butoxybenzenesulfonyl chloride
Molecular Weight (g/mol): 248.72
Molecular Formula: C10H13ClO3S
InChi Key: HGKWMUBXVMFXNC-UHFFFAOYSA-N
SMILES: CCCCOC1=CC=C(C=C1)S(Cl)(=O)=O
Quantity 
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (2)