accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1138011-22-9

IUPAC Name: 2,2,2-trifluoro-1-(pyridin-3-yl)ethan-1-amine hydrochloride
Synonyms: 2,2,2-trifluoro-1-(pyridin-3-yl)ethanamine hydrochloride
Molecular Weight (g/mol): 212.6
Molecular Formula: C7H8ClF3N2
InChi Key: NLDVAGWIUPSGQC-UHFFFAOYNA-N
SMILES: Cl.NC(C1=CN=CC=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results