accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3CCH3CH3CH3CH3CH3CCH3CH3CH3CH3CH3CCH3CH3CH3CH3CH3CCH3CH3ClClClClRuRuRuRu

CAS RN 113860-07-4

IUPAC Name: tetrakis(λ²-ruthenium(2+)) tetrakis(pentamethylcyclopenta-2,4-dien-1-ide) tetrachloride
Synonyms: tetrakis(λ²-ruthenium(2+)) tetrakis(pentamethylcyclopenta-2,4-dien-1-ide) tetrachloride
Molecular Weight (g/mol): 1087
Molecular Formula: C40H60Cl4Ru4
InChi Key: BCYJCIIJQVNGGB-UHFFFAOYSA-J
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Ru++].[Ru++].[Ru++].[Ru++].C[C-]1C(C)=C(C)C(C)=C1C.C[C-]1C(C)=C(C)C(C)=C1C.C[C-]1C(C)=C(C)C(C)=C1C.C[C-]1C(C)=C(C)C(C)=C1C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results