CAS RN 114-21-6
IUPAC Name:
sodium 2-hydroxy-2-phenylacetate
Synonyms:
sodium mandelate
Molecular Weight (g/mol):
174.13
Molecular Formula:
C8H7NaO3
InChi Key:
FWDLHTBMGQEUDU-UHFFFAOYNA-M
SMILES:
[Na+].OC(C([O-])=O)C1=CC=CC=C1
1
–
1
of
1
results
1
Sodium DL-Mandelate 99.0+%, TCI America™
CAS: 114-21-6 Molecular Formula: C8H7NaO3 Molecular Weight (g/mol): 174.131 MDL Number: MFCD00051122 InChI Key: FWDLHTBMGQEUDU-UHFFFAOYSA-M Synonym: DL-Mandelic Acid Sodium Salt PubChem CID: 23675514 IUPAC Name: sodium;2-hydroxy-2-phenylacetate SMILES: C1=CC=C(C=C1)C(C(=O)[O-])O.[Na+]