CAS RN 1140-14-3
IUPAC Name:
(2,4-dimethylphenyl)(phenyl)methanone
Synonyms:
(2,4-dimethylphenyl)(phenyl)methanone
Molecular Weight (g/mol):
210.28
Molecular Formula:
C15H14O
InChi Key:
UYSQHMXRROFKRN-UHFFFAOYSA-N
SMILES:
CC1=CC=C(C(=O)C2=CC=CC=C2)C(C)=C1
1
–
1
of
1
results
1
2,4-Dimethylbenzophenone 96.0+%, TCI America™
CAS: 1140-14-3 Molecular Formula: C15H14O Molecular Weight (g/mol): 210.276 MDL Number: MFCD00008537 InChI Key: UYSQHMXRROFKRN-UHFFFAOYSA-N PubChem CID: 70831 IUPAC Name: (2,4-dimethylphenyl)-phenylmethanone SMILES: CC1=CC(=C(C=C1)C(=O)C2=CC=CC=C2)C