accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNON

CAS RN 1141-06-6

IUPAC Name: 2-hydroxy-1,2-bis(pyridin-2-yl)ethan-1-one
Synonyms: 2-hydroxy-1,2-bis(pyridin-2-yl)ethanone
Molecular Weight (g/mol): 214.22
Molecular Formula: C12H10N2O2
InChi Key: ZKBDAJDDDOIASC-UHFFFAOYNA-N
SMILES: OC(C(=O)C1=CC=CC=N1)C1=CC=CC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results