accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1141474-53-4

IUPAC Name: 3-(1,3-dioxolan-2-yl)-N'-hydroxybenzene-1-carboximidamide
Synonyms: 3-(1,3-dioxolan-2-yl)-N'-hydroxybenzenecarboximidamide
Molecular Weight (g/mol): 208.22
Molecular Formula: C10H12N2O3
InChi Key: HOIMCQQBWIATEJ-UHFFFAOYSA-N
SMILES: NC(=NO)C1=CC(=CC=C1)C1OCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results