accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1141878-45-6

IUPAC Name: [1-(oxan-2-yl)-3-(trifluoromethyl)-1H-pyrazol-5-yl]boronic acid
Synonyms: 2-(oxan-2-yl)-5-(trifluoromethyl)pyrazol-3-ylboronic acid
Molecular Weight (g/mol): 264.01
Molecular Formula: C9H12BF3N2O3
InChi Key: IIRVABFYRULQMI-UHFFFAOYNA-N
SMILES: OB(O)C1=CC(=NN1C1CCCCO1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results