CAS RN 1142-20-7
N-Benzyloxycarbonyl-L-alanine, 98%
CAS: 1142-20-7 Molecular Formula: C11H13NO4 Molecular Weight (g/mol): 223.23 MDL Number: MFCD00002640 InChI Key: TYRGLVWXHJRKMT-QMMMGPOBSA-N Synonym: n-carbobenzyloxy-l-alanine,z-ala-oh,n-cbz-l-alanine,z-l-alanine,n-benzyloxycarbonyl-l-alanine,cbz-l-alanine,cbz-ala-oh,carbobenzyloxy-l-alanine,benzyloxycarbonyl-l-alanine,carbobenzoxy-l-alanine PubChem CID: 736104 SMILES: C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
Thermo Scientific Chemicals N-Carbobenzyloxy-L-alanine, 98%
CAS: 1142-20-7 Molecular Formula: C11H13NO4 Molecular Weight (g/mol): 223.23 MDL Number: MFCD00002640 InChI Key: TYRGLVWXHJRKMT-QMMMGPOBSA-N Synonym: n-carbobenzyloxy-l-alanine,z-ala-oh,n-cbz-l-alanine,z-l-alanine,n-benzyloxycarbonyl-l-alanine,cbz-l-alanine,cbz-ala-oh,carbobenzyloxy-l-alanine,benzyloxycarbonyl-l-alanine,carbobenzoxy-l-alanine PubChem CID: 736104 IUPAC Name: (2S)-2-(phenylmethoxycarbonylamino)propanoic acid SMILES: C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
N-Carbobenzoxy-L-alanine 99.0+%, TCI America™
CAS: 1142-20-7 Molecular Formula: C11H13NO4 Molecular Weight (g/mol): 223.23 MDL Number: MFCD00002640 InChI Key: TYRGLVWXHJRKMT-QMMMGPOBSA-N Synonym: n-carbobenzyloxy-l-alanine,z-ala-oh,n-cbz-l-alanine,z-l-alanine,n-benzyloxycarbonyl-l-alanine,cbz-l-alanine,cbz-ala-oh,carbobenzyloxy-l-alanine,benzyloxycarbonyl-l-alanine,carbobenzoxy-l-alanine PubChem CID: 736104 IUPAC Name: (2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid SMILES: C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
Z-Ala-OH, Advanced ChemTech
CAS: 1142-20-7 Molecular Formula: C11H13NO4 Molecular Weight (g/mol): 223.23 MDL Number: MFCD00002640 InChI Key: TYRGLVWXHJRKMT-QMMMGPOBSA-N IUPAC Name: (2S)-2-{[(benzyloxy)carbonyl]amino}propanoic acid SMILES: C[C@H](NC(=O)OCC1=CC=CC=C1)C(O)=O
Sigma Aldrich Boc-Phe-Osu
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.