accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1142363-56-1

IUPAC Name: 4,4,5,5-tetramethyl-2-(2,2,6,6-tetramethyl-3,6-dihydro-2H-pyran-4-yl)-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-(2,2,6,6-tetramethyl-3H-pyran-4-yl)-1,3,2-dioxaborolane
Molecular Weight (g/mol): 266.19
Molecular Formula: C15H27BO3
InChi Key: VOXXPMAJNSVUKJ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(C)(C)OC(C)(C)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results