accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3H3CCH3CH3BrH3CCH3H3C

CAS RN 1142818-90-3

IUPAC Name: 4-bromo-1,1,7,7-tetraethyl-3,3,5,5-tetramethyl-1,2,3,5,6,7-hexahydro-s-indacene
Synonyms: 4-bromo-1,1,7,7-tetraethyl-3,3,5,5-tetramethyl-2,6-dihydro-s-indacene
Molecular Weight (g/mol): 405.46
Molecular Formula: C24H37Br
InChi Key: DTVNULIVVOTHAI-UHFFFAOYSA-N
SMILES: CCC1(CC)CC(C)(C)C2=C(Br)C3=C(C=C12)C(CC)(CC)CC3(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results