accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrH2NN

CAS RN 114344-60-4

IUPAC Name: 2-amino-3-bromobenzonitrile
Synonyms: 2-amino-3-bromobenzonitrile
Molecular Weight (g/mol): 197.04
Molecular Formula: C7H5BrN2
InChi Key: RTIWACSVMFUEBF-UHFFFAOYSA-N
SMILES: NC1=C(C=CC=C1Br)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results