accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 114382-20-6

IUPAC Name: 5-(2-bromophenyl)-1H-pyrazole
Synonyms: 3-(2-bromophenyl)-2H-pyrazole
Molecular Weight (g/mol): 223.07
Molecular Formula: C9H7BrN2
InChi Key: VDHTYVJAQNZWTD-UHFFFAOYSA-N
SMILES: BrC1=CC=CC=C1C1=CC=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results