accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOHNOOOOClCl

CAS RN 1144035-53-9

IUPAC Name: (3,5-dichlorophenyl)methyl 4-[3-oxo-3-(2-oxo-2,3-dihydro-1,3-benzoxazol-6-yl)propyl]piperazine-1-carboxylate
Synonyms: (3,5-dichlorophenyl)methyl 4-[3-oxo-3-(2-oxo-3H-1,3-benzoxazol-6-yl)propyl]piperazine-1-carboxylate
Molecular Weight (g/mol): 478.33
Molecular Formula: C22H21Cl2N3O5
InChi Key: JMSUDQYHPSNBSN-UHFFFAOYSA-N
SMILES: ClC1=CC(COC(=O)N2CCN(CCC(=O)C3=CC=C4NC(=O)OC4=C3)CC2)=CC(Cl)=C1
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results