accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 114435-02-8

IUPAC Name: 4-fluoro-1,3-dioxolan-2-one
Synonyms: 4-fluoro-1,3-dioxolan-2-one
Molecular Weight (g/mol): 106.05
Molecular Formula: C3H3FO3
InChi Key: SBLRHMKNNHXPHG-UHFFFAOYNA-N
SMILES: FC1COC(=O)O1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)