accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOOHNaNaNa

CAS RN 114456-61-0

IUPAC Name: trisodium 2-hydroxypropane-1,2,3-tricarboxylate
Synonyms: trisodium citrate|citric acid trisodium salt
Molecular Weight (g/mol): 258.07
Molecular Formula: C6H5Na3O7
InChi Key: HRXKRNGNAMMEHJ-UHFFFAOYSA-K
SMILES: [Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Grade 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results