accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 1145-56-8

OHOHONHOCl

CAS RN 1145-56-8

IUPAC Name: 2-(2-chloroacetamido)-3-(4-hydroxyphenyl)propanoic acid
Synonyms: 2-(2-chloroacetamido)-3-(4-hydroxyphenyl)propanoic acid
Molecular Weight (g/mol): 257.67
Molecular Formula: C11H12ClNO4
InChi Key: GDOGSOZOUAVIFX-UHFFFAOYNA-N
SMILES: OC(=O)C(CC1=CC=C(O)C=C1)NC(=O)CCl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)