accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOO

CAS RN 1145-76-2

IUPAC Name: 1-(benzyloxy)-4-nitrobenzene
Synonyms: 1-(benzyloxy)-4-nitrobenzene
Molecular Weight (g/mol): 229.24
Molecular Formula: C13H11NO3
InChi Key: YOVUXLHIVNBVKO-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(OCC2=CC=CC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results