accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3ONHHN

CAS RN 114636-31-6

IUPAC Name: N-(pyrrolidin-3-yl)acetamide
Synonyms: N-(pyrrolidin-3-yl)acetamide
Molecular Weight (g/mol): 128.18
Molecular Formula: C6H12N2O
InChi Key: HDCCJUCOIKLZNM-UHFFFAOYNA-N
SMILES: CC(=O)NC1CCNC1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)