accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 114745-26-5

IUPAC Name: N-{9-[(2R,3R,4R,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxy-3-methoxyoxolan-2-yl]-6-oxo-6,9-dihydro-3H-purin-2-yl}-2-methylpropanamide
Synonyms: N-{9-[(2R,3R,4R,5R)-5-{[bis(4-methoxyphenyl)(phenyl)methoxy]methyl}-4-hydroxy-3-methoxyoxolan-2-yl]-6-oxo-3H-purin-2-yl}-2-methylpropanamide
Molecular Weight (g/mol): 669.74
Molecular Formula: C36H39N5O8
InChi Key: ISQLJOGRNUQHJX-QTXIGGTANA-N
SMILES: CO[C@@H]1[C@H](O)[C@@H](COC(C2=CC=CC=C2)(C2=CC=C(OC)C=C2)C2=CC=C(OC)C=C2)O[C@H]1N1C=NC2=C1NC(NC(=O)C(C)C)=NC2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results